Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 8 - 9 | 817 | 100 | |
NM_001307364.3 | NP_001294293.1 | 7 - 8 | 823 | 100 | ||
NM_001307365.3 | NP_001294294.1 | 6 - 7 | 700 | 100 | ||
NM_068737.2 | NP_501138.1 | 8 - 9 | 820 | 100 | ||
GenBank mRNA | U76402.1 | - | 8 - 9 | 820 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |