Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dif-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383120.1 | 7 - 8 | 3572 | 79 | ||
GenBank mRNA | X76115.1 | - | 310 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |