Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nep-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028032.6 | NP_001023203.2 | 14 - 15 | 2573 | 98 | |
NM_001028034.5 | NP_001023205.2 | 12 - 13 | 2237 | 98 | ||
NM_001383127.1 | 13 - 14 | 2369 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |