Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068878.5 | NP_501279.1 | 5 - 6 | 1907 | 75 | |
GenBank mRNA | AF072907.1 | - | 5 - 6 | 1902 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |