Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307234.3 | NP_001294163.1 | 1 - 2 | 139 | 79 | |
NM_001372874.1 | NP_001360383.1 | 4 - 5 | 737 | 79 | ||
NM_068885.8 | NP_501286.2 | 4 - 5 | 586 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |