Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068936.5 | NP_501337.1 | 2 - 3 | 246 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |