Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabs-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307243.2 | NP_001294172.1 | 4 - 5 | 665 | 78 | |
NM_068951.4 | NP_501352.1 | 4 - 5 | 667 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |