Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pct-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068971.7 | NP_501372.1 | 4 - 5 | 488 | 76 | |
GenBank mRNA | AF129110.1 | - | 3 - 4 | 434 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |