Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068998.10 | NP_501399.1 | 2 - 3 | 2151 | 131 | |
GenBank mRNA | AB055110.1 | - | 2 - 3 | 2173 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |