Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069046.6 | NP_501447.1 | 6 - 7 | 864 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |