Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307115.4 | NP_001294044.2 | 7 - 8 | 1532 | 108 | |
NM_001307116.3 | NP_001294045.1 | 5 - 6 | 1180 | 108 | ||
NM_001307117.3 | NP_001294046.1 | 5 - 6 | 1180 | 108 | ||
NM_001380369.1 | NP_001368458.1 | 5 - 6 | 982 | 108 | ||
NM_001380370.1 | NP_001368457.1 | 5 - 6 | 982 | 108 | ||
NM_069054.6 | NP_501455.2 | 7 - 8 | 1533 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |