Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069074.7 | NP_501475.1 | 9 - 10 | 3173 | 66 | |
GenBank mRNA | AF163018.1 | - | 9 - 10 | 3158 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |