Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-33E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069079.6 | NP_501480.2 | 2 - 3 | 291 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |