Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hst-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069090.5 | NP_501491.4 | 2 - 3 | 896 | 73 | |
GenBank mRNA | AB038044.1 | - | 2 - 3 | 916 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |