Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028199.5 | NP_001023370.1 | 7 - 8 | 836 | 86 | |
NM_001028200.6 | NP_001023371.1 | 8 - 9 | 1047 | 86 | ||
NM_001047522.4 | NP_001040987.1 | 8 - 9 | 1053 | 86 | ||
NM_001380387.1 | NP_001367079.1 | 8 - 9 | 1047 | 86 | ||
NM_069133.5 | NP_501534.2 | 8 - 9 | 1047 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |