Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268436.3 | NP_001255365.1 | 7 - 8 | 1283 | 157 | |
NR_056354.1 | 2 - 3 | 426 | 157 | |||
GenBank mRNA | X64962.1 | - | 676 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |