Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307015.3 | NP_001293944.1 | 6 - 7 | 1410 | 84 | |
NM_069279.4 | NP_501680.2 | 6 - 7 | 1425 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |