Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cka-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136377.4 | NP_001129849.1 | 9 - 10 | 1092 | 165 | |
NM_001306954.3 | NP_001293883.1 | 9 - 10 | 1103 | 165 | ||
NM_069331.4 | NP_501732.1 | 10 - 11 | 1170 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |