Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047489.4 | NP_001040954.1 | 3 - 4 | 466 | 66 | |
GenBank mRNA | AF332211.1 | - | 3 - 4 | 466 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |