Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380420.1 | NP_001366963.1 | 7 - 8 | 861 | 99 | |
GenBank mRNA | AF103732.1 | - | 7 - 8 | 856 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |