Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dur-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027974.4 | NP_001023145.1 | 15 - 16 | 1052 | 66 | |
NM_001268504.3 | NP_001255433.1 | 18 - 19 | 1247 | 66 | ||
NM_001268505.3 | NP_001255434.1 | 19 - 20 | 1313 | 66 | ||
NM_001268506.3 | NP_001255435.1 | 19 - 20 | 1316 | 66 | ||
NM_001268507.3 | NP_001255436.1 | 21 - 22 | 2150 | 66 | ||
NM_001268509.3 | NP_001255438.1 | 4 - 5 | 305 | 66 | ||
NM_001268510.3 | NP_001255439.1 | 12 - 13 | 830 | 66 | ||
NM_001268511.3 | NP_001255440.1 | 2 - 3 | 182 | 66 | ||
NM_001268514.3 | NP_001255443.1 | 8 - 9 | 587 | 66 | ||
NM_001307013.3 | NP_001293942.1 | 15 - 16 | 983 | 66 | ||
NM_001372910.1 | NP_001360167.1 | 13 - 14 | 899 | 66 | ||
NM_001383174.1 | 13 - 14 | 954 | 66 | |||
NM_069385.6 | NP_501786.2 | 19 - 20 | 2028 | 66 | ||
NM_069386.8 | NP_501787.3 | 20 - 21 | 2081 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |