Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uba-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038315.8 | NP_001033404.1 | 6 - 7 | 3229 | 121 | |
NM_001380424.1 | NP_001367213.1 | 6 - 7 | 3204 | 121 | ||
NM_001383176.1 | 6 - 7 | 3142 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |