Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069419.2 | NP_501820.2 | 6 - 7 | 893 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |