Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306925.2 | NP_001293854.1 | 6 - 7 | 659 | 83 | |
NM_001306926.3 | NP_001293855.1 | 3 - 4 | 422 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |