Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ech-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380437.1 | NP_001368477.1 | 7 - 8 | 1086 | 102 | |
NM_069474.6 | NP_501875.2 | 7 - 8 | 1141 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |