Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028294.4 | NP_001023465.1 | 5 - 6 | 691 | 78 | |
NM_001276842.3 | NP_001263771.1 | 3 - 4 | 592 | 78 | ||
NM_001383185.1 | 4 - 5 | 619 | 78 | |||
GenBank mRNA | AB011382.1 | - | 3 - 4 | 593 | 78 | |
AF061201.1 | - | 3 - 4 | 593 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |