Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268551.3 | NP_001255480.1 | 12 - 13 | 1377 | 76 | |
NM_001268552.3 | NP_001255481.1 | 1 - 2 | 132 | 76 | ||
NM_001372918.2 | NP_001359633.1 | 13 | 1438 | 76 | ||
NM_001380440.2 | NP_001366994.1 | 12 - 13 | 1452 | 76 | ||
GenBank mRNA | AF233263.1 | - | 9 - 10 | 1047 | 76 | |
AF233264.1 | - | 9 - 10 | 1047 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |