Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069554.8 | NP_501955.2 | 1 - 2 | 140 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |