Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069567.7 | NP_501968.2 | 3 - 4 | 533 | 73 | |
GenBank mRNA | AJ582070.1 | - | 2 - 3 | 422 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |