Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028110.4 | NP_001023281.1 | 14 - 15 | 10836 | 90 | |
NM_001028111.5 | NP_001023282.1 | 13 - 14 | 10728 | 90 | ||
NM_001268571.3 | NP_001255500.1 | 14 - 15 | 10767 | 90 | ||
NM_001268572.3 | NP_001255501.1 | 13 - 14 | 10707 | 90 | ||
NM_001307078.4 | NP_001294007.1 | 14 - 15 | 10807 | 90 | ||
NM_001307079.3 | NP_001294008.1 | 15 - 16 | 10857 | 90 | ||
GenBank mRNA | AB001074.1 | - | 14 - 15 | 10830 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |