Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: miz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268583.2 | NP_001255512.1 | 6 - 7 | 897 | 98 | |
NM_001268584.2 | NP_001255513.1 | 2 - 3 | 375 | 98 | ||
NM_001307057.4 | NP_001293986.1 | 6 - 7 | 899 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |