Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: miz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268583.2 | NP_001255512.1 | 8 - 9 | 1318 | 131 | |
NM_001268584.2 | NP_001255513.1 | 4 - 5 | 796 | 131 | ||
NM_001307057.4 | NP_001293986.1 | 8 - 9 | 1320 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |