Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306993.3 | NP_001293922.1 | 5 - 6 | 2949 | 110 | |
NM_069631.7 | NP_502032.2 | 5 - 6 | 2947 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |