Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268592.3 | NP_001255521.1 | 5 - 6 | 677 | 65 | |
NM_001383200.1 | 5 - 6 | 722 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |