Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069693.5 | NP_502094.1 | 4 - 5 | 456 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |