Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069696.5 | NP_502097.1 | 2 - 3 | 558 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |