Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028377.5 | NP_001023548.2 | 12 - 13 | 3076 | 107 | |
NM_001307645.3 | NP_001294574.1 | 12 - 13 | 3076 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |