Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067614.4 | NP_500015.3 | 5 - 6 | 679 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |