Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356908.3 | NP_001343633.1 | 4 | 646 | 67 | |
NM_001372721.2 | NP_001359917.1 | 3 - 4 | 552 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |