Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067710.7 | NP_500111.3 | 4 - 5 | 799 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |