Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067710.7 | NP_500111.3 | 6 - 7 | 1582 | 69 |
Gene Symbol: nhr-146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129338.5 | NP_001122810.1 | 1 - 2 | 77 | 69 |
Gene Symbol: nhr-87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383042.1 | 1 - 2 | 111 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |