Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067743.7 | NP_500144.2 | 5 - 6 | 890 | 90 | |
GenBank mRNA | AF353303.1 | - | 5 - 6 | 890 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |