Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307651.3 | NP_001294580.1 | 3 - 4 | 507 | 78 | |
NM_001356909.3 | NP_001343690.1 | 13 - 14 | 2382 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |