Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028280.5 | NP_001023451.1 | 2 - 3 | 865 | 74 | |
NM_001380087.1 | NP_001367641.1 | 1 - 2 | 166 | 74 | ||
GenBank mRNA | AB009567.1 | - | 1 - 2 | 188 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |