Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028280.5 | NP_001023451.1 | 4 - 5 | 1024 | 105 | |
GenBank mRNA | AB009567.1 | - | 3 - 4 | 347 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |