Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppgn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067790.7 | NP_500191.3 | 7 - 8 | 1287 | 87 | |
GenBank mRNA | AB257343.1 | - | 6 - 7 | 1237 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |