Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lad-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028325.3 | NP_001023496.1 | 14 - 15 | 2417 | 98 | |
GenBank mRNA | AF456361.1 | - | 14 - 15 | 2421 | 98 | |
EF584655.1 | - | 14 - 15 | 2331 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |