Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307537.3 | NP_001294466.1 | 10 - 11 | 9666 | 70 | |
NM_001307538.3 | NP_001294467.1 | 10 - 11 | 9603 | 70 | ||
NM_001307539.3 | NP_001294468.1 | 10 - 11 | 9675 | 70 | ||
NM_001307540.3 | NP_001294469.1 | 10 - 11 | 9525 | 70 | ||
NM_001307541.2 | NP_001294470.1 | 10 - 11 | 9660 | 70 | ||
NM_001307542.3 | NP_001294471.1 | 10 - 11 | 9534 | 70 | ||
NM_001307543.3 | NP_001294472.1 | 10 - 11 | 9669 | 70 | ||
NM_067883.5 | NP_500284.2 | 10 - 11 | 9658 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |