Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307537.3 | NP_001294466.1 | 11 - 12 | 9919 | 58 | |
NM_001307538.3 | NP_001294467.1 | 11 - 12 | 9856 | 58 | ||
NM_001307539.3 | NP_001294468.1 | 11 - 12 | 9928 | 58 | ||
NM_001307540.3 | NP_001294469.1 | 11 - 12 | 9778 | 58 | ||
NM_001307541.2 | NP_001294470.1 | 11 - 12 | 9913 | 58 | ||
NM_001307542.3 | NP_001294471.1 | 11 - 12 | 9787 | 58 | ||
NM_001307543.3 | NP_001294472.1 | 11 - 12 | 9922 | 58 | ||
NM_067883.5 | NP_500284.2 | 11 - 12 | 9911 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |