Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307538.3 | NP_001294467.1 | 2 - 3 | 710 | 76 | |
NM_001307540.3 | NP_001294469.1 | 2 - 3 | 641 | 76 | ||
NM_001307542.3 | NP_001294471.1 | 2 - 3 | 641 | 76 | ||
NM_067883.5 | NP_500284.2 | 2 - 3 | 774 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |